Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ACSL1 Antibodies

          Proteintech

          ACSL1 Polyclonal Antibody

          36 References
          View all (14) ACSL1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ACSL1 Polyclonal Antibody

          • Antibody Testing Data (16)
          ACSL1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ACSL1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ACSL1 Antibody (13989-1-AP) in ICC/IF

          Immunofluorescent analysis of ( 10% Formaldehyde ) fixed HepG2 cells using 13989-1-AP (ACSL1 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ACSL1 Antibody in Immunocytochemistry (ICC/IF)
          ACSL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACSL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACSL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACSL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACSL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACSL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACSL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACSL1 Antibody in Western Blot (WB)
          ACSL1 Antibody in Western Blot (WB)
          ACSL1 Antibody in Western Blot (WB)
          ACSL1 Antibody in Western Blot (WB)
          ACSL1 Antibody in Western Blot (WB)
          ACSL1 Antibody in Immunoprecipitation (IP)
          ACSL1 Antibody in Immunoprecipitation (IP)
          ACSL1 Antibody in Immunoprecipitation (IP)

          Product Details

          13989-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 29 publications 29 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:400
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication

          Miscellaneous PubMed (MISC)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ACSL1 Fusion Protein Ag5059 (349-698 aa encoded by BC050073)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.22 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          An anti-rat ACSL1 antibody recognized a band of the predicted 68 kDa in high-speed supernatant from rat liver and in human and murine SMCs, monocyte-derived macrophages, and murine peritoneal macrophages. It has 2 isoforms produced by alternative splicing.

          Immunogen sequence: RLLMDDLKV LQPTVFPVVP RLLNRMFDRI FGQANTTLKR WLLDFASKRK EAELRSGIIR NNSLWDRLIF HKVQSSLGGR VRLMVTGAAP VSATVLTFLR AALGCQFYEG YGQTECTAGC CLTMPGDWTA GHVGAPMPCN LIKLVDVEEM NYMAAEGEGE VCVKGPNVFQ GYLKDPAKTA EALDKDGWLH TGDIGKWLPN GTLKIIDRKK HIFKLAQGEY IAPEKIENIY MRSEPVAQVF VHGESLQAFL IAIVVPDVET LCSWAQKRGF EGSFEELCRN KDVKKAILED MVRLGKDSGL KPFEQVKGIT LHPELFSIDN GLLTPTMKAK RPELRNYFRS QIDDLYSTIK V (349-698 aa encoded by BC050073)

          Target Information

          The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ACS1; Acyl CoA synthetase, long chain; Acyl-CoA synthetase 1; Arachidonate--CoA ligase; fatty acid Coenzyme A ligase, long chain 2; fatty-acid-Coenzyme A ligase, long-chain 1; fatty-acid-Coenzyme A ligase, long-chain 2; L LACS2; LACS 1; LACS 2; lignoceroyl-CoA synthase; Long-chain acyl-CoA synthetase 1; Long-chain acyl-CoA synthetase 2; Long-chain fatty acid-CoA ligase 2; long-chain fatty-acid-coenzyme A ligase 1; Long-chain-fatty-acid--CoA ligase 1; Long-chain-fatty-acid--CoA ligase, liver isozyme; OTTHUMP00000218457; OTTHUMP00000218460; OTTHUMP00000218462; OTTHUMP00000218465; Palmitoyl-CoA ligase 1; Palmitoyl-CoA ligase 2; paltimoyl-CoA ligase 1; Phytanate--CoA ligase

          View more View less

          Gene Aliases: Acas; Acas1; ACS; ACS1; ACSL1; Acsl2; COAA; FACL1; FACL2; FACS; LACS; LACS 1; LACS1; LACS2

          View more View less

          UniProt ID: (Human) P33121, (Mouse) P41216, (Rat) P18163

          View more View less

          Entrez Gene ID: (Human) 2180, (Mouse) 14081, (Rat) 25288

          View more View less

          Function(s)
          long-chain fatty acid-CoA ligase activity ATP binding decanoate--CoA ligase activity acetate-CoA ligase activity
          Process(es)
          long-chain fatty acid metabolic process triglyceride metabolic process response to nutrient lipid biosynthetic process response to organic cyclic compound fatty acid transport adiponectin-activated signaling pathway response to oleic acid long-chain fatty-acyl-CoA biosynthetic process alpha-linolenic acid metabolic process xenobiotic catabolic process response to drug linoleic acid metabolic process long-chain fatty acid import positive regulation of protein serine/threonine kinase activity lipid metabolic process fatty acid metabolic process response to organic substance
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0