Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAVSNIRYG AAVTKEVGMD LKNMGAKNVC LMTDKNLSKL PPVQVAMDSL VKNGIPFTVY DNVRVEPTDS SFMEAIEFAQ KGAFDAYVAV GGGSTMDTCK AANLYASSPH SDFLDYVSAP IGKGKPVSVP LKPLIAVPTT SGTGSETTGV AIFDYEHLKV KIGITSRAIK PTLGLIDPLH TLHMPARVVA NSGFDVLCHA LESYTTLPYH LRSPCPSNPI TRPAYQGSNP ISDIWAIHAL RIVAKYLKRA VNSTDK (1-255 aa encoded by B C064634)
The ADHFE1 gene encodes hydroxyacid-oxoacid transhydrogenase (EC 1.1.99.24), which is responsible for the oxidation of 4-hydroxybutyrate in mammalian tissues (Kardon et al., 2006 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADHFe1; alcohol dehydrogenase 8; Alcohol dehydrogenase iron-containing protein 1; alcohol dehydrogenase, iron containing, 1; Fe-containing alcohol dehydrogenase; Fe-containing alcohol dehydrogenase 1; HOT; Hydroxyacid-oxoacid transhydrogenase, mitochondrial
Gene Aliases: 6330565B14Rik; ADH8; ADHFE1; AI043035; HMFT2263; HOT
UniProt ID: (Human) Q8IWW8, (Mouse) Q8R0N6, (Rat) Q4QQW3
Entrez Gene ID: (Human) 137872, (Mouse) 76187, (Rat) 362474
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support