Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-sheep serum, purified and partially purified Alkaline Phosphatase [Calf intestine]. Cross reactivity against Alkaline Phosphatase from other tissues and species may occur but have not been specifically determined. No reaction was observed against the bacterial form of the enzyme.
Alkaline phosphatase is a homodimeric protein enzyme (86 kDa), containing two zinc atoms crucial to its catalytic function per monomer, and it is optimally active at alkaline pH environments. The enzyme is found in prokaryotes and eukaryotes, with the same general function but in different structural forms suitable to organism's environment. ALP plays a role in liver metabolism and development within the skeleton. In bacteria, ALP acts as a source of inorganic phosphate. In yeast, ALP can be a useful protein for studying membrane protein sorting.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alkaline phosphatase liver/bone/kidney isozyme; alkaline phosphatase tissue non-specific isoform; Alkaline phosphatase, tissue-nonspecific isozyme; AP-TNAP; FLJ40094; FLJ93059; glycerophosphatase; liver/bone/kidney isozyme; MGC161443; MGC167935; Phosphoamidase; Phosphocreatine phosphatase; TNSALP
Gene Aliases: ALPL; BOS_2299; TNS-AP
UniProt ID: (Bovine) P09487
Entrez Gene ID: (Bovine) 280994
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support