Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Description: Intercellular Adhesion Molecule-3 (ICAM-3) is a member of the immunoglobulin supergene family and functions as a ligand for the Lymphocyte Function-Associated Antigen-1 (LFA-1). ICAM-3 is a heavily glycosylated, integral membrane protein with five immunoglobulin-like domains. It shares high homology to ICAM-1 and ICAM-2 in the extracellular region. In contrast to ICAM-1 and ICAM-2, ICAM-3 is absent on endothelia. ICAM-3 is expressed on resting lymphocytes, monocytes and neutrophils, representing the major LFA-1 ligand on these cells. The finding that adhesion of resting T lymphocytes to LFA-1 occurs primarily via ICAM-3 combined with the fact that ICAM-3 is much better expressed than other LFA-1 ligands on monocytes and resting lymphocytes implies an important role for ICAM-3 in the initiation of immune responses.
Applications Tested: ELISA.
CD50 (intracellular adhesion molecule 3, ICAM-3) is a transmembrane glycoprotein expressed by leukocytes, that serves as a counter-receptor for the lymphocyte function-associated antigen (LFA)-1 integrin. Besides functioning as an adhesive molecule that mediates e.g. the contact between T cells and antigen presenting cells, ICAM-3 regulates affinity of LFA-1 for ICAM-1 and induces T cell activation and proliferation. ICAM-3 plays an essential role in the initiation of the immune response both on T cells and antigen presenting cells and interacts also with CD209 (dendritic cell-specific ICAM-3-grabbing nonintegrin, DC-SIGN), a C-type lectin of dendritic cells and macrophages; this process is involved in dialogue between dendritic cells and granulocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD50; CDw50; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3
Gene Aliases: CD50; CDW50; ICAM-R; ICAM3
UniProt ID: (Human) P32942
Entrez Gene ID: (Human) 3385
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support