Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: EATFCDFPKI NHGILYDEEK YKPFSQVPTG EVFYYSCEYN FVSPSKSFWT RITCTEEGWS PTPKCLRLCF FPFVENGHSE SSGQTHLEGD TVQIICNTGY RLQNNENNIS CVERGWSTPP KCRSTDTSCV NPPTVQNAHI LSRQMSKYPS GERVRYECRS PYEMFGDEEV MCLNGNWTEP PQCKDSTGKC GPPPPIDNGD ITSFPLSVYA PASSVEYQCQ NLYQLEGNKR ITCRNGQWSE PPKCLHPCVI SREIMENYNI ALRWTAKQKL YLRTGESAEF VCKRGYRLSS RSHTLRTTCW DGKLEYPTCA KR
This gene encodes a secreted protein belonging to the complement factor H protein family. It binds to Pseudomonas aeruginosa elongation factor Tuf together with plasminogen, which is proteolytically activated. It is proposed that Tuf acts as a virulence factor by acquiring host proteins to the pathogen surface, controlling complement, and facilitating tissue invasion. Mutations in this gene are associated with an increased risk of atypical hemolytic-uremic syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: complement factor H-related 1 pseudogene; Complement factor H-related protein 1; FHL-1; FHR-1; H factor (complement)-like 1; H factor (complement)-like 2; H factor-like protein 1; H-factor-like 1; H36
Gene Aliases: CFHL; CFHL1; CFHL1P; CFHR1; CFHR1P; FHR1; H36-1; H36-2; HFL1; HFL2
UniProt ID: (Human) Q03591
Entrez Gene ID: (Human) 3078
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support