Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MPRYTVHVR GEWLAVPCQD AQLTVGWLGR EAVRRYIKNK PDNGGFTSVD DAHFLVRRCK GLGLLDNEDR LEVALENNEF VEVVIEGDAM SPDFIPSQPE GVYLYSKYRE PEKYIELDGD RLTTEDLVNL GKGRYKIKLT PTAEKRVQKS REVIDSIIKE KTVVYGITTG FGKFARTVIP INKLQELQVN LVRSHSSGVG KPLSPERCRM LLALRINVLA KGYSGISLET LKQVIEMFNA SCLPYVPEKG TVGASGDLAP LSHLALGLVG EGKMWSPKSG WADAKYVLEA HGLKPVILKP KEGLALINGT QMITSLGCEA VERASAIARQ ADIVAALTLE VLKGTTKAFD TDIHAL (1-355 aa encoded by B C096097)
HAL is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. HAL defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histidase; Histidine ammonia-lyase; histidinemia
Gene Aliases: HAL; HIS; histidase; Hsd; HSTD; Huth
UniProt ID: (Human) P42357, (Rat) P21213, (Mouse) P35492
Entrez Gene ID: (Human) 3034, (Rat) 29301, (Mouse) 15109
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support