Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Antibody detects endogenous levels of total IFNA5.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFN-alpha-5; Interferon; interferon alpha family, gene 5; Interferon alpha-5; Interferon alpha-61; Interferon alpha-G; interferon, alpha 5; LeIF G
Gene Aliases: Ifa5; IFN-alpha-5; IFN-alphaG; IFNA5; INA5; INFA5; leIF G
UniProt ID: (Human) P01569
Entrez Gene ID: (Human) 3442, (Rat) 690894, (Mouse) 15968
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support