Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SRC2 Antibodies

          Fabgennix

          KAT13C/NCOA2 Polyclonal Antibody, FITC

          View all (15) SRC2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite KAT13C/NCOA2 Polyclonal Antibody, FITC

          Product Details

          NUCR2-FITC

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1:10,000
          -

          Immunoprecipitation (IP)

          1:50-1:250
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide taken within amino acid 1375-1425 on rat NuCR2/GRIP-1/SRC-2 protein.
          View immunogen

          Conjugate

          FITC FITC FITC

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5-1.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          proprietary buffer, pH 7.4-7.8, with 30% glycerol, 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          SRC2 (steroid receptor coactivator 2), also known as nuclear receptor coactivator2, NCOA2 or TIF2, aids in the function of nuclear hormone receptors. Nuclear hormone receptors are conditional transcription factors that play important roles in various aspects of cell growth, development, and homeostasis by controlling expression of specific genes. Members of the nuclear hormone receptor superfamily, which includes the 5 steroid receptors and class II nuclear receptors, are structurally characterized by 3 distinct domains: an N-terminal transcriptional activation domain, a central DNA-binding domain, and a C-terminal hormone-binding domain. Before the binding of hormone, steroid receptors, which are sometimes called class I of the nuclear hormone receptor family, remain inactive in a complex with heat-shock protein-90 and other stress family proteins. Binding of hormone induces critical conformational changes in steroid receptors that cause them to dissociate from the inhibitory complex, bind as homodimers to specific DNA enhancer elements associated with target genes, and modulate that gene's transcription. After binding to enhancer elements, transcription factors require transcriptional coactivator proteins to mediate their stimulation of transcription initiation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: bHLHe75; c-fgr; c-src2; Class E basic helix-loop-helix protein 75; FLJ43153; Glucocorticoid receptor-interacting protein 1; glucocorticoid receptor-interacting protein-1; GRIP-1; hTIF2; MGC75096; NCoA-2; Nuclear receptor coactivator 2; p160 steroid receptor coactivator 2; p55c-fgr; p58c-fgr; SRC-2; Steroid receptor coactivator 2; TIF 2; Transcriptional intermediary factor 2

          View more View less

          Gene Aliases: BHLHE75; D1Ertd433e; GRIP-1; GRIP1; KAT13C; NCoA-2; NCOA2; SRC-2; SRC2; TIF2

          View more View less

          UniProt ID: (Human) Q15596, (Mouse) Q61026, (Rat) Q9WUI9

          View more View less

          Entrez Gene ID: (Human) 10499, (Mouse) 17978, (Rat) 83724

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding chromatin binding transcription coactivator activity histone acetyltransferase activity protein binding ligand-dependent nuclear receptor binding PDZ domain binding estrogen receptor binding ligand-dependent nuclear receptor transcription coactivator activity thyroid hormone receptor coactivator activity protein complex binding progesterone receptor binding nuclear hormone receptor binding glucocorticoid receptor binding retinoid X receptor binding thyroid hormone receptor binding protein dimerization activity DNA polymerase binding signal transducer activity receptor binding transcription factor binding retinoic acid receptor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated male gonad development regulation of glucose metabolic process bile acid and bile salt transport histone acetylation cerebellum development intracellular receptor signaling pathway cellular response to hormone stimulus circadian regulation of gene expression cellular lipid metabolic process locomotor rhythm positive regulation of female receptivity positive regulation of transcription from RNA polymerase II promoter cellular response to Thyroglobulin triiodothyronine positive regulation of receptor activity positive regulation of glucocorticoid receptor signaling pathway signal transduction negative regulation of transcription, DNA-templated
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline