Product References
GIPC3 couples to MYO6 and PDZ domain proteins and shapes the hair cell apical region.
bioRxiv : the preprint server for biology
Chatterjee P,Morgan CP,Krey JF,Benson C,Goldsmith J,Bateschell M,Ricci AJ,Barr-Gillespie PG
Wed Mar 01 00:00:00 UTC 2023
GIPC3 couples to MYO6 and PDZ domain proteins, and shapes the hair cell apical region.
Journal of cell science
Chatterjee P,Morgan CP,Krey JF,Benson C,Goldsmith J,Bateschell M,Ricci AJ,Barr-Gillespie PG
Mon May 15 00:00:00 UTC 2023
The Smad4-MYO18A-PP1A complex regulates β-catenin phosphorylation and pemigatinib resistance by inhibiting PAK1 in cholangiocarcinoma.
Cell death and differentiation
Liu J,Ren G,Li K,Liu Z,Wang Y,Chen T,Mu W,Yang X,Li X,Shi A,Zhao W,Xu B,Chang J,Guo S,Pan C,Zhou T,Zhang Z,Xu Y
Fri Apr 01 00:00:00 UTC 2022
The Smad4-MYO18A-PP1A complex regulates β-catenin phosphorylation and pemigatinib resistance by inhibiting PAK1 in cholangiocarcinoma.
Cell death and differentiation
Liu J,Ren G,Li K,Liu Z,Wang Y,Chen T,Mu W,Yang X,Li X,Shi A,Zhao W,Xu B,Chang J,Guo S,Pan C,Zhou T,Zhang Z,Xu Y
Fri Apr 01 00:00:00 UTC 2022
A protumorigenic secretory pathway activated by p53 deficiency in lung adenocarcinoma.
The Journal of clinical investigation
Tan X,Shi L,Banerjee P,Liu X,Guo HF,Yu J,Bota-Rabassedas N,Rodriguez BL,Gibbons DL,Russell WK,Creighton CJ,Kurie JM
Mon Jan 04 00:00:00 UTC 2021
Genomic and epigenomic adaptation in SP-R210 (Myo18A) isoform-deficient macrophages.
Immunobiology
Yau E,Chen Y,Song C,Webb J,Carillo M,Kawasawa YI,Tang Z,Takahashi Y,Umstead TM,Dovat S,Chroneos ZC
Mon Nov 01 00:00:00 UTC 2021
Unbiased Profiling of the Human Proinsulin Biosynthetic Interaction Network Reveals a Role for Peroxiredoxin 4 in Proinsulin Folding.
Diabetes
Tran DT,Pottekat A,Mir SA,Loguercio S,Jang I,Campos AR,Scully KM,Lahmy R,Liu M,Arvan P,Balch WE,Kaufman RJ,Itkin-Ansari P
Sat Aug 01 00:00:00 UTC 2020
Pushing myelination - developmental regulation of myosin expression drives oligodendrocyte morphological differentiation.
Journal of cell science
Domingues HS,Urbanski MM,Macedo-Ribeiro S,Almaktari A,Irfan A,Hernandez Y,Wang H,Relvas JB,Rubinstein B,Melendez-Vasquez CV,Pinto IM
Wed Aug 05 00:00:00 UTC 2020
APOL1 C-Terminal Variants May Trigger Kidney Disease through Interference with APOL3 Control of Actomyosin.
Cell reports
Uzureau S,Lecordier L,Uzureau P,Hennig D,Graversen JH,Homblé F,Mfutu PE,Oliveira Arcolino F,Ramos AR,La Rovere RM,Luyten T,Vermeersch M,Tebabi P,Dieu M,Cuypers B,Deborggraeve S,Rabant M,Legendre C,Moestrup SK,Levtchenko E,Bultynck G,Erneux C,Pérez-Morga D,Pays E
Tue Mar 17 00:00:00 UTC 2020
A novel isoform of myosin 18A (Myo18Aγ) is an essential sarcomeric protein in mouse heart.
The Journal of biological chemistry
Horsthemke M,Nutter LMJ,Bachg AC,Skryabin BV,Honnert U,Zobel T,Bogdan S,Stoll M,Seidl MD,Müller FU,Ravens U,Unger A,Linke WA,van Gorp PRR,de Vries AAF,Bähler M,Hanley PJ
Fri May 03 00:00:00 UTC 2019
A Portrait of the Human Organelle Proteome In Space and Time during Cytomegalovirus Infection.
Cell systems
Jean Beltran PM,Mathias RA,Cristea IM
Wed Oct 26 00:00:00 UTC 2016
A Portrait of the Human Organelle Proteome In Space and Time during Cytomegalovirus Infection.
Cell systems
Jean Beltran PM,Mathias RA,Cristea IM
Wed Oct 26 00:00:00 UTC 2016
A Portrait of the Human Organelle Proteome In Space and Time during Cytomegalovirus Infection.
Cell systems
Jean Beltran PM,Mathias RA,Cristea IM
Wed Oct 26 00:00:00 UTC 2016
A Portrait of the Human Organelle Proteome In Space and Time during Cytomegalovirus Infection.
Cell systems
Jean Beltran PM,Mathias RA,Cristea IM
Wed Oct 26 00:00:00 UTC 2016
Identification of CD245 as myosin 18A, a receptor for surfactant A: A novel pathway for activating human NK lymphocytes.
Oncoimmunology
De Masson A,Giustiniani J,Marie-Cardine A,Bouaziz JD,Dulphy N,Gossot D,Validire P,Tazi A,Garbar C,Bagot M,Merrouche Y,Bensussan A
Sun May 01 00:00:00 UTC 2016
Identification of CD245 as myosin 18A, a receptor for surfactant A: A novel pathway for activating human NK lymphocytes.
Oncoimmunology
De Masson A,Giustiniani J,Marie-Cardine A,Bouaziz JD,Dulphy N,Gossot D,Validire P,Tazi A,Garbar C,Bagot M,Merrouche Y,Bensussan A
Sun May 01 00:00:00 UTC 2016
GM-CSF modulates pulmonary resistance to influenza A infection.
Antiviral research
Sever-Chroneos Z,Murthy A,Davis J,Florence JM,Kurdowska A,Krupa A,Tichelaar JW,White MR,Hartshorn KL,Kobzik L,Whitsett JA,Chroneos ZC
Tue Nov 01 00:00:00 UTC 2011
GM-CSF modulates pulmonary resistance to influenza A infection.
Antiviral research
Sever-Chroneos Z,Murthy A,Davis J,Florence JM,Kurdowska A,Krupa A,Tichelaar JW,White MR,Hartshorn KL,Kobzik L,Whitsett JA,Chroneos ZC
Tue Nov 01 00:00:00 UTC 2011
Surfactant protein A (SP-A)-mediated clearance of Staphylococcus aureus involves binding of SP-A to the staphylococcal adhesin eap and the macrophage receptors SP-A receptor 210 and scavenger receptor class A.
The Journal of biological chemistry
Sever-Chroneos Z,Krupa A,Davis J,Hasan M,Yang CH,Szeliga J,Herrmann M,Hussain M,Geisbrecht BV,Kobzik L,Chroneos ZC
Fri Feb 11 00:00:00 UTC 2011
Surfactant protein A (SP-A)-mediated clearance of Staphylococcus aureus involves binding of SP-A to the staphylococcal adhesin eap and the macrophage receptors SP-A receptor 210 and scavenger receptor class A.
The Journal of biological chemistry
Sever-Chroneos Z,Krupa A,Davis J,Hasan M,Yang CH,Szeliga J,Herrmann M,Hussain M,Geisbrecht BV,Kobzik L,Chroneos ZC
Fri Feb 11 00:00:00 UTC 2011