Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAEADFKMVS EPVAHGVAEE EMASSTSDSG EESDSSSSSS STSDSSSSSS TSGSSSGSGS SSSSSGSTSS RSRLYRKKRV PEPSRRARRA PLGTNFVDRL PQAVRNRVQA LRNIQDECDK VDTLFLKAIH DLERKYAELN KPLYDRRFQI INAEYEPTEE ECEWNSEDEE FSSDEEVQDN TPSEMPPLEG EEEENPKENP EVKAEEKEVP KEIPEVKDEE KEVAKEITEV KAEEKADSKD CMEATPEVKE DPKEVPQVKA DDKEQPKATE AKARAAVRET HKRVPEERLQ DSVDLKRARK GKPKREDPKG IPDYWLIVLK NVDKLGPMIQ KYDEPILKFL SDVSLKFSKP GQPVSYTFEF HFLPNPYFRN EVLVKTYIIK AKPDHNDPFF SWGWEIEDCK GCKIDWRRGK DVTVTTTQSR TTATGEIEIQ PRVVPNASFF NFFSPPEIPM IGKLEPREDA ILDEDFEIGQ ILHDNVILKS IYYYTGEVNG TYYQFGKHYG NKKYRK
This gene is intronless and encodes a member of the nucleosome assembly protein (NAP) family. This gene is linked closely to a region of genes responsible for several X-linked mental retardation syndromes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nucleosome assembly protein 1-like 3
Gene Aliases: BNAP; MB20; NAP1L3; NPL3
UniProt ID: (Human) Q99457
Entrez Gene ID: (Human) 4675
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support