Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDLWSAGCVF YEIASLQPLF PGVNELDQIS KIHDVIGTPA QKILTKFKQS RAMNFDFPFK KGSGIPLLTT NLSPQCLSLL HAMVAYDPDE RIAAHQALQH PYFQEQRKTE KRALGSHRKA GFPEHPVAPE PLSNSCQISK EGRKQKQSLK QEEDRPKRRG PAYVMELPKL KLSGVVRLSS YSSPTLQSVL GSGTNGRVPV LRPLKCIPAS KKTDPQKDLK PAPQQCRLPT IVRKGGR
MOK is able to phosphorylate several exogenous substrates and to undergo autophosphorylation. It negatively regulates cilium length in a cAMP and mTORC1 signaling-dependent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAPK/MAK/MRK overlapping kinase; MOK protein kinase; RAGE-1; renal cell carcinoma antigen; Renal tumor antigen 1; soluble RAGE
Gene Aliases: MOK; RAGE; RAGE-1; RAGE1; STK30
UniProt ID: (Human) Q9UQ07
Entrez Gene ID: (Human) 5891
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support