Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: EGIRDKVSDW DEFLRQTLIG ACSPPVPLLE GLRNGRNPLD LIAPGSRLEC QAFQDSLSTW IVTVVENIGG RLKLRYEGLE SSDNYEHWLY YLDPFLHHVG WAAQQGYELQ
This gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. It encodes a protein which contains four malignant brain tumor repeat (mbt) domains and may be involved in antigen recognition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSFMBT; Renal ubiquitous protein 1; Scm-like with four MBT domains protein 1; Scm-related gene containing four mbt domains; Scm-related gene product containing four mbt domains
Gene Aliases: hSFMBT; RU1; SFMBT; SFMBT1
UniProt ID: (Human) Q9UHJ3
Entrez Gene ID: (Human) 51460
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support