Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: WTSTHGGSG SRQGRQDGDQ DAPDIRNRFP NFLGIIRLYD YLQQLHENPQ SLGTYLQGTA SALSQSQEPP PSVNRVPPSS PSSQEPGSGQ PLPEESVAIK GRSSCPAFLR YPTENSTGQG GDQDGAGKSS TGHSTNLPDL VSGLGDSANR VPFAPLSFSP TAAIPGIPEP PWLPSPAYPR SLRPDGMNPA PQLQDEIQPQ LPLLMHLQAA MANPRALQAL RQIEQGLQVL ATEAPRLLLW FMPCLAGTGS VAGGIESRED PLMSEDPLPN PPPEVFPALD SAELGFLSPP FLHMLQDLVS TNPQQLQPEA HFQVQL (306-620 aa encoded by BC036743)
This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This gene is specifically expressed in the testis, and proposed to regulate cell-cycle progression during spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: testicular tissue protein Li 220; Ubiquilin-3
Gene Aliases: 4933400K24Rik; TUP-1; UBQLN3
UniProt ID: (Human) Q9H347, (Mouse) Q8C5U9
Entrez Gene ID: (Human) 50613, (Mouse) 244178
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support