Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: SSALKIEIYP TSLTKTELEN FIMQQIKRWS DLPKDTFWLY FSQVQEIMSI FGDITRKFCK DVVSNKKFMK KVQESRFDVI FADAIFPCS
The UGTs serve a major role in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. UGT2B7 has unique specificity for 3,4-catechol estrogens and estriol, suggesting that it may play an important role in regulating the level and activity of these potent estrogen metabolites. Its subcellular location is the microsome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3,4-catechol estrogen specific; 3,4-catechol estrogen-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B7; UDP glucuronosyltransferase 2B7; UDP-glucuronosyltransferase 2B7; UDP-glucuronosyltransferase 2B8; UDP-glucuronosyltransferase 2B9; UDP-glucuronyltransferase, family 2, beta-7; UDPGT; UDPGT 2B7; UDPGT 2B8; UDPGT 2B9; UDPGTh 2; UDPGTh-2; UGT2B-RH4
Gene Aliases: UDPGT 2B7; UDPGT 2B9; UDPGT2B7; UDPGTH2; UGT2B7; Ugt2b8; UGT2B9; UGTB2B9
UniProt ID: (Human) P16662, (Rat) Q62789
Entrez Gene ID: (Human) 7364, (Rat) 286989
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support