Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Sequence of this protein is as follows: MPGKMVVILG ASNILWIMFA ASQAFKIETT PESRYLAQIG DSVSLTCSTT GCESPFFSWR TQIDSPLNGK VTNEGTTSTL TMNPVSFGNE HSYLCTATCE SRKLEKGIQV EIYSFPKDPE IHLSGPLEAG KPITVKCSVA DVYPFDRLEI DLLKGDHLMK SQEFLEDADR KSLETKSLEV TFTPVIEDIG KVLVCRAKLH IDEMDSVPTV RQAVKELQVY ISPKNTVISV NPSTKLQEGG SVTMTCSSEG LPAPEIFWSK KLDNGNLQHL SGNATLTLIA MRMEDSGIYV CEGVNLIGKN RKEVELIVQE KPFTVEISPG PRIAAQIGDS VMLTCSVMGC ESPSFSWRTQ IDSPLSGKVR SEGTNSTLTL SPVSFENEHS YLCTVTCGHK KLEKGIQVEL YSFPRDPEIE MSGGLVNGSS VTVSCKVPSV YPLDRLEIEL LKGETILENI EFLEDTDMKS LENKSLEMTF IPTIEDTGKA LVCQAKLHID DMEFEPKQRQ STQTLYVNVA PRDTTVLVSP SSILEEGSSV NMTCLSQGFP APKILWSRQL PNGELQPLSE NATLTLISTK MEDSGVYLCE GINQAGRSRK EVELIIQVTP KDIKLTAFPS ESVKEGDTVI ISCTCGNVPE TWIILKKKAE TGDTVLKSID GAYTIRKAQL KDAGVYECES KNKVGSQLRS LTLDVQGREN NKDYFSPELL VLYFASSLII PAIGMIIYFA RKANMKGSYS LVEAQKSKV
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD106; CD106 antigen; DKFZp779G2333; INCAM-100; INCAM100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; V-CAM 1; Vascular cell adhesion protein 1
Gene Aliases: CD106; INCAM-100; Vcam-1; VCAM1
UniProt ID: (Human) P19320
Entrez Gene ID: (Human) 7412, (Mouse) 22329
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support