Search Thermo Fisher Scientific
Progen
Specificity: the p27 subunit of the 20S subcomplex within the 26S hetero-oligomeric protein complex and the free cytosolic form of 20S cylinder particles. Polypeptide reacting: p27 subunit of 20S cylinder particles.Reactivity on cultured cell lines: XLKE-A6 (Xenopus laevis), PLC (human), RVF (rat), Ptk2 (rat, kangaroo).