Product References
Integrin α6 signaling induces STAT3-TET3-mediated hydroxymethylation of genes critical for maintenance of glioma stem cells.
Oncogene
Herrmann A,Lahtz C,Song J,Aftabizadeh M,Cherryholmes GA,Xin H,Adamus T,Lee H,Grunert D,Armstrong B,Chu P,Brown C,Lim M,Forman S,Yu H
Research topic: Cancer, Glioma, stem cells
Sun Mar 01 00:00:00 UTC 2020
Small-Molecule Targeting of Oncogenic FTO Demethylase in Acute Myeloid Leukemia.
Cancer cell
Huang Y,Su R,Sheng Y,Dong L,Dong Z,Xu H,Ni T,Zhang ZS,Zhang T,Li C,Han L,Zhu Z,Lian F,Wei J,Deng Q,Wang Y,Wunderlich M,Gao Z,Pan G,Zhong D,Zhou H,Zhang N,Gan J,Jiang H,Mulloy JC,Qian Z,Chen J,Yang CG
Research topic: acute myeloid leukemia (AML)
Mon Apr 15 00:00:00 UTC 2019
Global DNA Methylation and Histone Posttranslational Modifications in Human and Nonhuman Primate Brain in Association with Prenatal Alcohol Exposure.
Alcoholism, clinical and experimental research
Jarmasz JS,Stirton H,Basalah D,Davie JR,Clarren SK,Astley SJ,Del Bigio MR
Research topic: Neurobiology, Alcoholism
Sat Jun 01 00:00:00 UTC 2019
Global DNA Methylation and Histone Posttranslational Modifications in Human and Nonhuman Primate Brain in Association with Prenatal Alcohol Exposure.
Alcoholism, clinical and experimental research
Jarmasz JS,Stirton H,Basalah D,Davie JR,Clarren SK,Astley SJ,Del Bigio MR
Research topic: Neurobiology, Alcoholism
Sat Jun 01 00:00:00 UTC 2019
Analysis of diet-induced differential methylation, expression, and interactions of lncRNA and protein-coding genes in mouse liver.
Scientific reports
Silva JP,van Booven D
Research topic: Liver
Wed Aug 01 00:00:00 UTC 2018
PTP1B controls non-mitochondrial oxygen consumption by regulating RNF213 to promote tumour survival during hypoxia.
Nature cell biology
Banh RS,Iorio C,Marcotte R,Xu Y,Cojocari D,Rahman AA,Pawling J,Zhang W,Sinha A,Rose CM,Isasa M,Zhang S,Wu R,Virtanen C,Hitomi T,Habu T,Sidhu SS,Koizumi A,Wilkins SE,Kislinger T,Gygi SP,Schofield CJ,Dennis JW,Wouters BG,Neel BG
Research topic: Hypoxia
Fri Jul 01 00:00:00 UTC 2016
DNA Hypomethylation and Histone Variant macroH2A1 Synergistically Attenuate Chemotherapy-Induced Senescence to Promote Hepatocellular Carcinoma Progression.
Cancer research
Borghesan M,Fusilli C,Rappa F,Panebianco C,Rizzo G,Oben JA,Mazzoccoli G,Faulkes C,Pata I,Agodi A,Rezaee F,Minogue S,Warren A,Peterson A,Sedivy JM,Douet J,Buschbeck M,Cappello F,Mazza T,Pazienza V,Vinciguerra M
Research topic: Liver Cancer
Mon Feb 01 00:00:00 UTC 2016
Tet1 and Tet2 Protect DNA Methylation Canyons against Hypermethylation.
Molecular and cellular biology
Wiehle L,Raddatz G,Musch T,Dawlaty MM,Jaenisch R,Lyko F,Breiling A
Research topic: DNA Methylation
Mon Feb 01 00:00:00 UTC 2016
Iodine-125 irradiation inhibits invasion of gastric cancer cells by reactivating microRNA-181c expression.
Oncology letters
Yang Y,Ma ZH,Li XG,Zhang WF,Wan J,Du LJ,Li GJ,Yang GK,Lu P
Research topic: Gastric Cancer
Sat Oct 01 00:00:00 UTC 2016
TET1 Depletion Induces Aberrant CpG Methylation in Colorectal Cancer Cells.
PloS one
Kai M,Niinuma T,Kitajima H,Yamamoto E,Harada T,Aoki H,Maruyama R,Toyota M,Sasaki Y,Sugai T,Tokino T,Nakase H,Suzuki H
Research topic: Colorectal
Wed Jul 05 00:00:00 UTC 2017
Paternal under-nutrition programs metabolic syndrome in offspring which can be reversed by antioxidant/vitamin food fortification in fathers.
Scientific reports
McPherson NO,Fullston T,Kang WX,Sandeman LY,Corbett MA,Owens JA,Lane M
Research topic: Metabolism
Fri Jun 03 00:00:00 UTC 2016
Hemi-methylated DNA regulates DNA methylation inheritance through allosteric activation of H3 ubiquitylation by UHRF1.
eLife
Harrison JS,Cornett EM,Goldfarb D,DaRosa PA,Li ZM,Yan F,Dickson BM,Guo AH,Cantu DV,Kaustov L,Brown PJ,Arrowsmith CH,Erie DA,Major MB,Klevit RE,Krajewski K,Kuhlman B,Strahl BD,Rothbart SB
Research topic: Epigenetic Inheritance
Tue Sep 06 00:00:00 UTC 2016
AF9 promotes hESC neural differentiation through recruiting TET2 to neurodevelopmental gene loci for methylcytosine hydroxylation.
Cell discovery
Qiao Y,Wang X,Wang R,Li Y,Yu F,Yang X,Song L,Xu G,Chin YE,Jing N
Research topic: Brain & Nerves
Wed Jul 27 00:00:00 UTC 2016
Genome-Wide Mapping of 5mC and 5hmC Identified Differentially Modified Genomic Regions in Late-Onset Severe Preeclampsia: A Pilot Study.
PloS one
Zhu L,Lv R,Kong L,Cheng H,Lan F,Li X
Research topic: Preeclampsia
Fri May 06 00:00:00 UTC 2016
Alteration of genic 5-hydroxymethylcytosine patterning in olfactory neurons correlates with changes in gene expression and cell identity.
Proceedings of the National Academy of Sciences of the United States of America
Colquitt BM,Allen WE,Barnea G,Lomvardas S
Research topic: Neurobiology
Tue Sep 03 00:00:00 UTC 2013