Antibody tested in indirect ELISA
6-Benzylaminopurine (6-BAP) is a synthetic cytokinin plant growth regulator that is commonly used in plant tissue culture and agricultural practices. It acts as a hormone to promote cell division and growth, leading to increased yield and quality of crops. 6-BAP has been shown to improve the rooting and shoot formation of plant cuttings, as well as increase fruit set and size in various crops. It is also used in the production of ornamental plants and for the rejuvenation of aging plants.