Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASMLLAQRL ACSFQHSYRL LVPGSRHISQ AAAKVDVEFD YDGPLMKTEV PGPRSQELMK QLNIIQNAEA VHFFCNYEES RGNYLVDVDG NRMLDLYSQI SSVPIGYSHP ALLKLIQQPQ NASMFVNRPA LGILPPENFV EKLRQSLLSV APKGMSQLIT MACGSCSNEN ALKTIFMWYR SKERGQRGFS QEELETCMIN QAPGCPDYSI LSFMGAFHGR TMGCLATTHS KAIHKIDIPS FDWPIAPFPR LKYPLEEFVK ENQQEEARCL EEVEDLIVKY RKKKKTVAGI IVEPIQSEGG DNHASDDFFR KLRDIARKHG CAFLVDEVQT GGGCTGKFWA HEHWGLDDPA DVMTFSKKMM TGGFFHKEEF RPNAPYRIFN TWLGDPSKNL LLAEVINIIK REDLLNNAAH AGKALLTGLL DLQARYPQFI SRVRGRGTFC SFDTPDDSIR NKLILIARNK GVVLGGCGDK SIRFRPTLVF RDHHAHLFLN IFSDILADFK
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (S)-3-amino-2-methylpropionate transaminase; 4-aminobutyrate aminotransferase, mitochondrial; 4-aminobutyrate transaminase; FLJ17813; FLJ30272; GABA aminotransferase; GABA AT; GABA T; GABA transaminase; GABA transferase; GABA-AT; Gamma-amino-N-butyrate transaminase; hCG1984265; L AIBAT; L-AIBAT
Gene Aliases: ABAT; GABA-AT; GABAT; NPD009
UniProt ID: (Human) P80404
Entrez Gene ID: (Human) 18
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support