Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ABHD5 Antibodies

          Proteintech

          ABHD5 Polyclonal Antibody

          26 References
          View all (13) ABHD5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ABHD5 Polyclonal Antibody

          • Antibody Testing Data (16)
          ABHD5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ABHD5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ABHD5 Antibody (12201-1-AP) in ICC/IF

          Immunofluorescent analysis of MCF-7 cells, using ABHD5 antibody 12201-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ABHD5 Antibody in Immunocytochemistry (ICC/IF)
          ABHD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ABHD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ABHD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ABHD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ABHD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ABHD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ABHD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ABHD5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ABHD5 Antibody in Western Blot (WB)
          ABHD5 Antibody in Western Blot (WB)
          ABHD5 Antibody in Western Blot (WB)
          ABHD5 Antibody in Western Blot (WB)
          ABHD5 Antibody in Western Blot (WB)
          ABHD5 Antibody in Western Blot (WB)
          ABHD5 Antibody in Immunoprecipitation (IP)

          Product Details

          12201-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 20 publications 20 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse, Pig

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ABHD5 Fusion Protein Ag2840 (6-349 aa encoded by BC021958)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: EEVDSADTG ERSGWLTGWL PTWCPTSISH LKEAEEKMLK CVPCTYKKEP VRISNGNKIW TLKFSHNISN KTPLVLLHGF GGGLGLWALN FGDLCTNRPV YAFDLLGFGR SSRPRFDSDA EEVENQFVES IEEWRCALGL DKMILLGHNL GGFLAAAYSL KYPSRVNHLI LVEPWGFPER PDLADQDRPI PVWIRALGAA LTPFNPLAGL RIAGPFGLSL VQRLRPDFKR KYSSMFEDDT VTEYIYHCNV QTPSGETAFK NMTIPYGWAK RPMLQRIGKM HPDIPVSVIF GARSCIDGNS GTSIQSLRPH SYVKTIAILG AGHYVYADQP EEFNQKVKEI CDTVD (6-349 aa encoded by BC021958)

          Target Information

          The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; Abhydrolase domain-containing protein 5; aCGI58; CGI 58; CGI58 protein; Lipid droplet-binding protein CGI-58; MGC8731; Protein CGI-58; truncated abhydrolase domain-containing protein 5

          View more View less

          Gene Aliases: 1300003D03Rik; 2010002J10Rik; ABHD5; CDS; CGI-58; CGI58; IECN2; IECN5; NCIE2

          View more View less

          UniProt ID: (Human) Q8WTS1, (Mouse) Q9DBL9

          View more View less

          Entrez Gene ID: (Human) 51099, (Mouse) 67469

          View more View less

          Function(s)
          1-acylglycerol-3-phosphate O-acyltransferase activity triglyceride lipase activity lysophosphatidic acid acyltransferase activity carboxylic ester hydrolase activity protein binding transferase activity transferase activity, transferring acyl groups
          Process(es)
          lipid metabolic process fatty acid metabolic process phosphatidic acid biosynthetic process phospholipid biosynthetic process negative regulation of sequestering of triglyceride positive regulation of triglyceride catabolic process cell differentiation positive regulation of lipoprotein lipase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0