Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The LIM domain is a double zinc finger structure that promotes protein-protein interactions. LIM domain proteins, such as ABLIM3, play roles in embryonic development, cell lineage determination, and cancer (Krupp et al., 2006 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: abLIM-3; actin binding LIM protein family, member 3; Actin-binding LIM protein 3; Actin-binding LIM protein family member 3
Gene Aliases: ABLIM3; AW990719; D930036B08Rik; HMFN1661; KIAA0843; RGD1565118
UniProt ID: (Human) O94929, (Mouse) Q69ZX8
Entrez Gene ID: (Human) 22885, (Rat) 307395, (Mouse) 319713
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support