Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEAEESEKAA TEQEPLEGTE QTLDAEEEQE ESEEAACGSK KRVVPGIVYL GHIPPRFRPL HVRNLLSAYG EVGRVFFQAE DRFVRRKKKA AAAAGGKKRS YTKDYTEGWV EFRDKRIAKR VAASLHNTPM GARRRSPFRY DLWNLKYLHR FTWSHLSEHL AFERQVRRQR LRAEVAQAKR ETDFYLQSVE RGQRFLAADG DPARPDGSWT FAQRPTEQEL RARKAARPGG RERARLATAQ DKARSNKGLL ARIFGAPPPS ESMEGPSLVR DS
Could be a novel TATA-binding protein (TBP) which can function as a basal transcription activator. Can act as a regulator of basal transcription for class II genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator of basal transcription 1; Basal transcriptional activator; hABT1; TATA-binding protein-binding protein
Gene Aliases: ABT1; hABT1
UniProt ID: (Human) Q9ULW3
Entrez Gene ID: (Human) 29777
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support