Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MQPEEGTGW LLELLSEVQL QQYFLRLRDD LNVTRLSHFE YVKNEDLEKI GMGRPGQRRL WEAVKRRKAL CKRKSWMSKV FSGKRLEAEF PPHHSQSTFR KTSPAPGGPA GEGPLQSLTC LIGEKDLRLL EKLGDGSFGV VRRGEWDAPS GKTVSVAVKC LKPDVLSQPE AMDDFIREVN AMHSLDHRNL IRLYGVVLTP PMKMVTELAP LGSLLDRLRK HQGHFLLGTL SRYAVQVAEG MGYLESKRFI HRDLAARNLL LATRDLVKIG DFGLMRALPQ NDDHYVMQEH RKVPFAWCAP ESLKTRTFSH ASDTWMFGVT LWEMFTYGQE PWIGLNGSQI LHKIDKEGER LPRPEDCPQD IYNVMVQCWA HKPEDRPTFV ALRDFL (1-385 aa encoded by B C028164)
TNK2 (ACK) (activated Cdc42-associated kinase) is a non-receptor tyrosine kinase that signals downstream of Cdc42. TNK2 is known to be involved in tumorigenesis and metastasis, particularly in prostate cancer. Downstream effector of CDC42 which mediates CDC42-dependent cell migration via phosphorylation of BCAR1. May be involved both in adult synaptic function and plasticity and in brain development. Activates AKT1 by phosphorylating it on 'Tyr-176'. Phosphorylates AR on 'Tyr-267' and 'Tyr-363' thereby promoting its recruitment to androgen-responsive enhancers (AREs). Phosphorylates WWOX on 'Tyr-287'. Phosphorylates MCF2, thereby enhancing its activity as a guanine nucleotide exchange factor (GEF) toward Rho family proteins. Contributes to the control of AXL receptor levels. Confers metastatic properties on cancer cells and promotes tumor growth by negatively regulating tumor suppressor such as WWOX and positively regulating pro-survival factors such as AKT1 and AR. Phosphorylates WASP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACK-1; Activated CDC42 kinase 1; activated Cdc42-associated kinase 1; activated p21cdc42Hs kinase; Cdc42 GTPase-inhibiting protein; Non-receptor protein tyrosine kinase Ack; P21cdc42Hs kinase; tnk2 {ECO:0000250|UniProtKB:Q07912}; Tyrosine kinase non-receptor protein 2; tyrosine kinase, non-receptor, 2
Gene Aliases: ACK; ACK-1; ACK1; Cdgip; p21cdc42Hs; Pyk1; TNK2
UniProt ID: (Human) Q07912, (Mouse) O54967, (Rat) Q5U2X5
Entrez Gene ID: (Human) 10188, (Mouse) 51789, (Rat) 303882
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support