Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ACOT7 Antibodies

          Proteintech

          ACOT7 Monoclonal Antibody (1E9C7)

          View all (6) ACOT7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ACOT7 Monoclonal Antibody (1E9C7)

          • Antibody Testing Data (1)
          ACOT7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ACOT7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ACOT7 Antibody (68140-1-IG) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with 68140-1-Ig (ACOT7 antibody) at dilution of 1:20000 incubated at room temperature for 1.5 hours.The membrane was stripped and reblotted with HRP-conjugated Alpha Tubulin Monoclonal antibody (HRP-66031) as loading control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ACOT7 Antibody in Western Blot (WB)

          Product Details

          68140-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rabbit, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1E9C7

          Immunogen

          Recombinant protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Protein A

          Storage buffer

          PBS with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: PDTCALLQPP AASAAAAPSM SGPDVETPSA IQICRIMRPD DANVAGNVHG GTILKMIEEA GAIISTRHCN SQNGERCVAA LARVERTDFL SPMCIGEVAH VSAEITYTSK HSVEVQVNVM SENILTGAKK LTNKATLWYV PLSLKNVDKV LEVPPVVYSR QEQEEEGRKR YEAQKLERME TKWRNGDIVQ PVLNPEPNTV SYSQSSLIHL VGPSDCTLHG FVHGGVTMKL MDEVAGIVAA RHCKTNIVTA SVDAINFHDK IRKGCVITIS GRMTFTSNKS MEIEVLVDAD PVVDSSQKRY RAASAFFTYV SLSQEGRSLP VPQLVPETED EKKRFEEGKG RYLQMKAKRQ GHAEPQP

          Target Information

          This gene encodes a member of the acyl coenzyme family. The encoded protein hydrolyzes the CoA thioester of palmitoyl-CoA and other long-chain fatty acids. Decreased expression of this gene may be associated with mesial temporal lobe epilepsy. Alternatively spliced transcript variants encoding distinct isoforms with different subcellular locations have been characterized.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 50-kDa brain acyl-CoA hydrolase; ACH1; ACT; acyl-CoA thioesterase 2; Acyl-CoA thioesterase 7; acyl-CoA thioesterase, long chain; BACH; brain acyl CoA hydrolase; Brain acyl-CoA hydrolase; CTE-II; CTE-IIa; CTE-IIb; Cytosolic acyl coenzyme A thioester hydrolase; L RP1-120G22.10; LACH1; Long chain acyl-CoA thioester hydrolase; MTE-II

          View more View less

          Gene Aliases: 2410041A17Rik; ACH1; ACOT7; ACT; BACH; CTE-II; CTE-IIa; hBACH; LACH; LACH1

          View more View less

          UniProt ID: (Human) O00154, (Mouse) Q91V12, (Rat) Q64559

          View more View less

          Entrez Gene ID: (Human) 11332, (Mouse) 70025, (Rat) 26759, (Rabbit) 100354701

          View more View less

          Function(s)
          fatty-acyl-CoA binding protein binding palmitoyl-CoA hydrolase activity long-chain fatty acyl-CoA binding protein homodimerization activity acyl-CoA hydrolase activity carboxylic ester hydrolase activity hydrolase activity
          Process(es)
          metabolic process fatty acid catabolic process coenzyme A biosynthetic process medium-chain fatty-acyl-CoA catabolic process long-chain fatty-acyl-CoA catabolic process medium-chain fatty acid biosynthetic process palmitic acid biosynthetic process long-chain fatty-acyl-CoA biosynthetic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0