Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Sequence of this protein is as follows: MDMWTALLIL QALLLPSLAD GATPALRFVA VGDWGGVPNA PFHTAREMAN AKEIARTVQI LGADFILSLG DNFYFTGVQD INDKRFQETF EDVFSDRSLR KVPWYVLAGN HDHLGNVSAQ IAYSKISKRW NFPSPFYRLH FKIPQTNVSV AIFMLDTVTL CGNSDDFLSQ QPERPRDVKL ARTQLSWLKK QLAAAREDYV LVAGHYPVWS IAEHGPTHCL VKQLRPLLAT YGVTAYLCGH DHNLQYLQDE NGVGYVLSGA GNFMDPSKRH QRKVPNGYLR FHYGTEDSLG GFAYVEISSK EMTVTYIEAS GKSLFKTRLP RRARP
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: human purple acid phosphatase; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; Tartrate-resistant acid phosphatase type 5; TR-AP; TRAPase; TrATPase; Type 5 acid phosphatase
Gene Aliases: ACP5; HPAP; TRACP5a; TRACP5b; TRAP; TrATPase
UniProt ID: (Human) P13686
Entrez Gene ID: (Human) 54
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support