Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNQIEPGVQY NYVYDEDEYM IQEEEWDRDL LLDPAWEKQQ RKTFTAWCNS HLRKAGTQIE NIEEDFRNGL KLMLLLEVIS GERLPKPDRG KMRFHKIANV NKALDYIASK GVKLVSIGAE EIVDGNVKMT LGMIWTIILR FAIQDISVEE TSAKEGLLLW CQRKTAPYRN VNIQNFHTSW KDGLGLCALI HRHRPDLIDY SKLNKDDPIG NINLAMEIAE KHLDIPKMLD AEDIVNTPKP DERAIMTYVS CFYHAFAGAE QAETAANRIC KVLAVNQENE RLMEEYERLA SELLEWIRRT IPWLENRTPE KTMQAMQKKL EDFRDYRRKH KPPKVQEKCQ LEINFNTLQT KLRISNRPAF MPSEGKMVSD IAGAWQRLEQ AEKGYEEWLL NEIRRLERLE HLAEKFRQKA STHETWAYGK EQILLQKDYE SASLTEVRAL LRKHEAFESD LAAHQDRVEQ IAAIAQELNE LDYHDAVNVN DRCQKICDQW DRLGTLTQKR REALERMEKL LETIDQLHLE FAKRAAPFNN WMEGAMEDLQ DMFIVHSIEE IQSLITAHEQ FKATLPEADG ERQSIMAIQN EVEKVIQSYN IRISSSNPYS TVTMDELRTK WDKVKQLVPI RDQSLQEELA RQHANERLRR QFAAQANAIG PWIQNKMEEI ARSSIQITGA LEDQMNQLKQ YEHNIINYKN NIDKLEGDHQ LIQEALVFDN KHTNYTMEHI RVGWELLLTT IARTINEVET QILTRDAKGI TQEQMNEFRA SFNHFDRRKN GLMDHEDFRA CLISMGYDLG EAEFARIMTL VDPNGQGTVT FQSFIDFMTR ETADTDTAEQ VIASFRILAS DKPYILAEEL RRELPPDQAQ YCIKRMPAYS GPGSVPGALD YAAFSSALYG ESDL
Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it i involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actinin, alpha 2; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin-2; F-actin cross-linking protein
Gene Aliases: ACTN2; CMD1AA; CMH23
UniProt ID: (Human) P35609
Entrez Gene ID: (Human) 88
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support