Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • alpha Actinin 4 Antibodies

          Proteintech

          ACTN4 Polyclonal Antibody

          22 References
          View all (34) alpha Actinin 4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ACTN4 Polyclonal Antibody

          • Antibody Testing Data (22)
          ACTN4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ACTN4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ACTN4 Antibody (19096-1-AP) in ICC/IF

          Immunofluorescent analysis of MCF-7 cells, using ACTN4 antibody 19096-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). Blue pseudocolor = DAPI (fluorescent DNA dye). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ACTN4 Antibody in Immunocytochemistry (ICC/IF)
          ACTN4 Antibody in Immunocytochemistry (ICC/IF)
          ACTN4 Antibody in Immunocytochemistry (ICC/IF)
          ACTN4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACTN4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACTN4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACTN4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACTN4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACTN4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Western Blot (WB)
          ACTN4 Antibody in Flow Cytometry (Flow)
          ACTN4 Antibody in Immunoprecipitation (IP)

          Product Details

          19096-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:8,000
          View 12 publications 12 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 5 publications 5 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ACTN4 Fusion Protein Ag13546 (612-911 aa encoded by BC005033)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.41 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody recognizes 100-105 kDa of endogenous ACTN4 protein in various lysates.

          Immunogen sequence: TTVTPQIIN SKWEKVQQLV PKRDHALLEE QSKQQSNEHL RRQFASQANV VGPWIQTKME EIGRISIEMN GTLEDQLSHL KQYERSIVDY KPNLDLLEQQ HQLIQEALIF DNKHTNYTME HIRVGWEQLL TTIARTINEV ENQILTRDAK GISQEQMQEF RASFNHFDKD HGGALGPEEF KACLISLGYD VENDRQGEAE FNRIMSLVDP NHSGLVTFQA FIDFMSRETT DTDTADQVIA SFKVLAGDKN FITAEELRRE LPPDQAEYCI ARMAPYQGPD AVPGALDYKS FSTALYGESD L (612-911 aa encoded by BC005033)

          Target Information

          Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, alpha actinin isoform which is concentrated in the cytoplasm, and thought to be involved in metastatic processes. Mutations in this gene have been associated with focal and segmental glomerulosclerosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: actinin-Alpha-4; alpha actinin 4; Alpha-actinin-4; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; Non-muscle alpha-actinin 4

          View more View less

          Gene Aliases: ACTININ-4; ACTN4; C77391; FSGS; FSGS1

          View more View less

          UniProt ID: (Human) O43707, (Mouse) P57780, (Rat) Q9QXQ0

          View more View less

          Entrez Gene ID: (Human) 81, (Mouse) 60595, (Rat) 63836

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding nucleoside binding actin binding integrin binding calcium ion binding protein binding ligand-dependent nuclear receptor transcription coactivator activity chromatin DNA binding nuclear hormone receptor binding protein homodimerization activity retinoic acid receptor binding ion channel binding poly(A) RNA binding protein N-terminus binding actin filament binding protein complex binding metal ion binding
          Process(es)
          transport protein transport peroxisome proliferator activated receptor signaling pathway regulation of apoptotic process retinoic acid receptor signaling pathway positive regulation of pinocytosis actin filament bundle assembly negative regulation of cellular component movement positive regulation of cellular component movement bicellular tight junction assembly protein localization to bicellular tight junction regulation of nucleic acid-templated transcription response to hypoxia platelet degranulation vesicle transport along actin filament positive regulation of cell migration positive regulation of sodium:proton antiporter activity negative regulation of substrate adhesion-dependent cell spreading positive regulation of NIK/NF-kappaB signaling
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0