Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ACVRL1 Antibodies

          Abnova

          ACVRL1 Monoclonal Antibody (7H2)

          View all (28) ACVRL1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ACVRL1 Monoclonal Antibody (7H2)

          • Antibody Testing Data (1)
          ACVRL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ACVRL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ACVRL1 Antibody (H00000094-M02) in WB

          Western Blot analysis of ACVRL1 expression in transfected 293T cell line by ACVRL1 monoclonal antibody (M02), clone 7H2.

          Lane 1: ACVRL1 transfected lysate(56.1 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ACVRL1 Antibody in Western Blot (WB)

          Product Details

          H00000094-M02

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          7H2

          Immunogen

          ACVRL1 (AAH42637, 22 a.a. approximately 119 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: DPVKPSRGPL VTCTCESPHC KGPTCRGAWC TVVLVREEGR HPQEHRGCGN LHRELCRGRP TEFVNHYCCD SHLCNHNVSL VLEATQPPSE QPGTDGQL

          Target Information

          Activin A receptor type II-like 1 (ACVRL1) is a type I cell-surface receptor for the TGF-beta superfamily of ligands. ACVRL1, similar to other type 1 receptors, has a conserved serine-threonine kinase subdomain, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. Mutations in ACVRL1 are associated with hemorrhagic telangiectasia type 2. Mutations in ACVRL1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like kinase 1; Activin receptor type-1-like; Activin receptor-like kinase 1; ALK-1; Serine/threonine-protein kinase receptor R3; SKR3; TGF-B superfamily receptor type I; TSR-I

          View more View less

          Gene Aliases: ACVRL1; ACVRLK1; ALK-1; ALK1; HHT; HHT2; ORW2; SKR3; TSR-I

          View more View less

          UniProt ID: (Human) P37023

          View more View less

          Entrez Gene ID: (Human) 94

          View more View less

          Function(s)
          protein serine/threonine kinase activity transmembrane receptor protein serine/threonine kinase activity receptor signaling protein serine/threonine kinase activity transforming growth factor beta-activated receptor activity transforming growth factor beta receptor activity, type I protein binding ATP binding activin receptor activity, type I protein kinase binding SMAD binding metal ion binding activin binding transforming growth factor beta binding BMP receptor activity
          Process(es)
          angiogenesis in utero embryonic development regulation of endothelial cell proliferation negative regulation of endothelial cell proliferation positive regulation of endothelial cell proliferation lymphangiogenesis blood vessel maturation blood vessel remodeling blood vessel endothelial cell proliferation involved in sprouting angiogenesis endocardial cushion morphogenesis regulation of DNA replication regulation of transcription, DNA-templated protein phosphorylation negative regulation of cell adhesion signal transduction transforming growth factor beta receptor signaling pathway blood circulation regulation of blood pressure negative regulation of cell proliferation negative regulation of endothelial cell migration negative regulation of gene expression positive regulation of pathway-restricted SMAD protein phosphorylation signal transduction by protein phosphorylation negative regulation of cell growth negative regulation of cell migration BMP signaling pathway positive regulation of BMP signaling pathway positive regulation of chondrocyte differentiation activin receptor signaling pathway wound healing, spreading of epidermal cells dorsal aorta morphogenesis regulation of blood vessel endothelial cell migration negative regulation of blood vessel endothelial cell migration negative regulation of endothelial cell differentiation positive regulation of endothelial cell differentiation positive regulation of angiogenesis positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter protein heterooligomerization negative regulation of focal adhesion assembly lymphatic endothelial cell differentiation artery development venous blood vessel development endothelial tube morphogenesis retina vasculature development in camera-type eye cellular response to transforming growth factor beta stimulus cellular response to BMP stimulus endocardial cushion to mesenchymal transition negative regulation of DNA biosynthetic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0