Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The ADPGK gene encodes ADP-dependent glucokinase, an enzyme distinct from the ATP-dependent hexokinases as it employs adenosine diphosphate (ADP) rather than ATP to phosphorylate glucose to glucose-6-phosphate. This gene, initially identified in archaeal species, has been co-opted into mammalian systems where it plays roles primarily in glycolytic processes. Structurally, the mammalian ADPGK differs from its archaeal counterpart, exemplified by the rearrangement of its secondary structures, such as the alpha2 and alpha3 helices. These helices are replaced by an extended beta-strand region, indicating evolutionary adaptations for function in higher organisms like mice. Although its precise physiological role remains partially defined, ADPGK is notably expressed in both hematopoietic lineages and tumor tissues, suggesting potential significance in cancer biology and energy metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-dependent glucokinase; ADP-GK; ATP-dependent glucokinase; EC 2.7.1.147; RbBP-35
Gene Aliases: 2610017G09Rik; ADP-GK; ADPGK; PSEC0260
UniProt ID: (Human) Q9BRR6
Entrez Gene ID: (Human) 83440
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support