Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: ASNKYLVEFR AGKMSLKGTT VTPDKRKGLV YIQQTDDSLI HFCWKDRTSG NVEDDLIIFP DDCEFKRVPQ CPSGRVYVLK FKAGSKRLFF WMQEPKTDQD EEHCRKVNEY LNNPPMPGAL GASGSSGHEL SALGGEGGLQ SLLGNMSHSQ LMQLIGPAGL GGLGGLGALT GPGLASLLGS SGPPGSSSSS SSRSQSAAVT PSSTTSSTRA TPAPSAPAAA SATSPSPAPS SGNGASTAAS PTQPIQLSDL QSILATMNVP AGPAGGQQVD LASVLAPEIM APILANADVQ ERLLPYLPSG ESLPQTADEI QNTLTSPQFQ QALGMFSAAL ASGQLGPLMC QFGLPAEAVE AANKGDVEAF AKAMQNNAKP EQKEGDTKDK KDEEEDMSLD
The protein encoded by this gene is an integral plasma membrane protein which promotes cell adhesion. The encoded protein is thought to undergo O-linked glycosylation. Expression of this gene has been shown to be induced by gamma interferon in some cancer cells. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 110 kDa cell membrane glycoprotein; Adhesion-regulating molecule 1; ARM-1; Gp110; hRpn13; M(r) 110,000 surface antigen; MGC29536; Proteasomal ubiquitin receptor ADRM1; Proteasome regulatory particle non-ATPase 13; Rpn13 homolog
Gene Aliases: ADRM1; ARM-1; ARM1; GP110
UniProt ID: (Human) Q16186
Entrez Gene ID: (Human) 11047
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support