Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • AFG3L2 Antibodies

          Abnova

          AFG3L2 Polyclonal Antibody, MaxPab™

          View all (16) AFG3L2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite AFG3L2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          AFG3L2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          AFG3L2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AFG3L2 Antibody (H00010939-B01P) in WB

          Western Blot analysis of AFG3L2 expression in transfected 293T cell line (H00010939-T01) by AFG3L2 MaxPab polyclonal antibody.

          Lane 1: AFG3L2 transfected lysate(87.67 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AFG3L2 Antibody in Western Blot (WB)

          Product Details

          H00010939-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          AFG3L2 (AAH65016.1, 1 a.a. approximately 797 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MAHRCLRLWG RGGCWPRGLQ QLLVPGGVGP GEQPCLRTLY RFVTTQARAS RNSLLTDIIA AYQRFCSRPP KGFEKYFPNG KNGKKASEPK EVMGEKKESK PAATTRSSGG GGGGGGKRGG KKDDSHWWSR FQKGDIPWDD KDFRMFFLWT ALFWGGVMFY LLLKRSGREI TWKDFVNNYL SKGVVDRLEV VNKRFVRVTF TPGKTPVDGQ YVWFNIGSVD TFERNLETLQ QELGIEGENR VPVVYIAESD GSFLLSMLPT VLIIAFLLYT IRRGPAGIGR TGRGMGGLFS VGETTAKVLK DEIDVKFKDV AGCEEAKLEI MEFVNFLKNP KQYQDLGAKI PKGAILTGPP GTGKTLLAKA TAGEANVPFI TVSGSEFLEM FVGVGPARVR DLFALARKNA PCILFIDEID AVGRKRGRGN FGGQSEQENT LNQLLVEMDG FNTTTNVVIL AGTNRPDILD PALLRPGRFD RQIFIGPPDI KGRASIFKVH LRPLKLDSTL EKDKLARKLA SLTPGFSGAD VANVCNEAAL IAARHLSDSI NQKHFEQAIE RVIGGLEKKT QVLQPEEKKT VAYHEAGHAV AGWYLEHADP LLKVSIIPRG KGLGYAQYLP KEQYLYTKEQ LLDRMCMTLG GRVSEEIFFG RITTGAQDDL RKVTQSAYAQ IVQFGMNEKV GQISFDLPRQ GDMVLEKPYS EATARLIDDE VRILINDAYK RTVALLTEKK ADVEKVALLL LEKEVLDKND MVELLGPRPF AEKSTYEEFV EGTGSLDEDT SLPEGLKDWN KEREKEKEEP PGEKVAN

          Target Information

          AFG3L2 is a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. AFG3L2 gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders. This gene encodes a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. This gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: AFG3 ATPase family gene 3-like 2; AFG3 ATPase family member 3-like 2; AFG3 like AAA ATPase 2; AFG3-like AAA ATPase 2; AFG3-like protein 2; ATPase family gene 3, yeast; FLJ25993; Mitochondrial inner membrane m-AAA protease component AFG3L2; Paraplegin-like protein

          View more View less

          Gene Aliases: AFG3L2; SCA28; SPAX5

          View more View less

          UniProt ID: (Human) Q9Y4W6

          View more View less

          Entrez Gene ID: (Human) 10939

          View more View less

          Function(s)
          ATP-dependent peptidase activity metalloendopeptidase activity protein binding ATP binding metallopeptidase activity zinc ion binding unfolded protein binding
          Process(es)
          protein complex assembly axonogenesis neuromuscular junction development mitochondrial fusion nerve development mitochondrial protein processing regulation of multicellular organism growth cristae formation myelination protein import into mitochondrial intermembrane space muscle fiber development righting reflex
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0