Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MFPALETHLK QTIPDPYEDF MYRHLQYYGY FKAQRGSLPN SATHQHVRKN NPQCLLNGSL GEKDDLIPDT LQKEKLLWPI SLSSAVHRQI EAINRDFHSC LGWMQWRGLS SLQPPPPRFK DSPASAFRVA GITDSHMLSL PHLRSRQLLY DELDEVNPRL REPQELFSIL STKRPLQAPR WPIECEVIKE NIHHIEWAPP QPEYFYQPKG NEKVPEIVGE KKGTVVYQLD SVPIEGSYFT SSRVGGKRGI VKELAVTLQG PEDNTLLFES RFESGNLQKA VRVDTYEYEL TLRTDLYTNK HTQWFYFRVQ NTRKDATYRF TIVNLLKPKS LYTVGMKPLL YSQLDANTRN IGWRREGNEI KYYKNNTDDG QQPFYCLTWT IQFPYDQDTC FFAHFYPYTY TDLQCYLLSV ANNPIQSQFC KLQTLCRSLA GNTVYLLTIT NPSQTPQEAA AKKAVVLSAR VHPGESNGSW VMKGFLDFIL SNSPDAQLLR DIFVFKVLPM LNPDGVIVGN YRCSLAGRDL NRHYKTILKE SFPCIWYTRN MIKRLLEERE VLLYCDFHGH SRKNNIFLYG CNNNNRKYWL HERVFPLMLC KNAPDKFSFH SCNFKVQKCK EGTGRVVMWR MGILNSYTME STFGGSTLGN KRDTHFTIED LKSLGYHVCD TLLDFCDPDQ IKFTQCLAEL KELLRQEIHK KFHELGQDVD LEGSWSDISL SDIESSTSGS DSSLSDGLPV HLANIADELT QKKKMFKKKK KKSLQTRKQR NEQYQKKNLM QKLKLTEDTS EKAGFASTLQ KQPTFFKNSE NSSFLPMKNE NPRLNETNLN RRDKDTPLDP SMATLILPKN KGRMQNKKPG FTVSCSPKRT INSSQEPAPG MKPNWPRSRY PATKRGCAAM AAYPSLHIYT YP
Metallocarboxypeptidase that mediates deglutamylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes gene-encoded polyglutamates from the carboxy-terminus of target proteins such as MYLK. Does not show detyrosinase or deglycylase activities from the carboxy-terminus of tubulin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP/GTP binding protein-like 2; ATP/GTP-binding protein-like 2; CBCP2; cytoplasmic carboxypeptidase 2; Cytosolic carboxypeptidase 2; Protein deglutamylase CCP2; testis tissue sperm-binding protein Li 96mP
Gene Aliases: AGBL2; CCP2
UniProt ID: (Human) Q5U5Z8
Entrez Gene ID: (Human) 79841
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support