Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMDWNEASSA PGYNWNQSVL FQSSSKPGRG RRKKVDLFEA SHLGFPTSAS AAASGYPSKR STGPRQPRGG RGGGACSAKK ERGGAAAKAK FIPKPQPVNP LFQDSPDLGL DYYSGDSSMS PLPSQSRAFG VGERDPCDFI GPYSMNPSTP SDGTFGQGFH CDSPSLGAPE LDGKHFPPLA HPPTVFDAGL QKAYSPTCSP TLGFKEELRP PPTKLAACEP LKHGLQGASL GHAAAAQAHL SCRDLPLGQP HYDSPSCKGT AYWYPPGSAA RSPPYEGKVG TGLLADFLGR TEAACLSAPH LASPPATPKA DKEPLEMARP PGPPRGPAAA AAGYGCPLLS DLTLSPVPRD SLLPLQDTAY RYPGFMPQAH PGLGGGPKSG FLGPMAEPHP EDTFTVTSL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AT hook, DNA binding motif, containing 1; AT-hook DNA-binding motif-containing protein 1; Transcription factor Gibbin
Gene Aliases: AHDC1; MRD25
UniProt ID: (Human) Q5TGY3
Entrez Gene ID: (Human) 27245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support