Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MRLPETQVL PGEIDETPLS KPGHDLASME DKTEKWSSQP EGPLKLKASS TDMPSQISVV NVDQLWEDSV LTVKFPKLMV PRFSFAAPSS EDDVFIPTVR EVQCPEANID TALCKESPGL WGASILKAGA GVPGE (1-134 aa encoded by BC049216)
The specific function of AHNAK2 is not yet known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein AHNAK2
Gene Aliases: AHNAK2; C14orf78; KIAA2019
UniProt ID: (Human) Q8IVF2
Entrez Gene ID: (Human) 113146
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support