Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSRQLSRARP ATVLGAMEMG RRMDAPTSAA VTRAFLERGH TEIDTAFVYS EGQSETILGG LGLRLGGSDC RVKIDTKAIP LFGNSLKPDS LRFQLETSLK RLQCPRVDLF YLHMPDHSTP VEETLRACHQ LHQEGKFVEL GLSNYAAWEV AEICTLCKSN GWILPTVYQG MYNAITRQVE TELFPCLRHF GLRFYAFNPL AGGLLTGKYK YEDKDGKQPV GRFFGNTWAE MYRNRYWKEH HFEGIALVEK ALQAAYGASA PSMTSATLRW MYHHSQLQGA HGDAVILGMS SLEQLEQNLA AAEEGPLEPA VVDAFNQAWH LVAHECPNYF R
Aldo-keto reductases, such as AKR7A3, are involved in the detoxification of aldehydes and ketones.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AFB1 aldehyde reductase 2; AFB1-AR 2; aflatoxin aldehyde reductase; aflatoxin B1 aldehyde reductase 2; Aflatoxin B1 aldehyde reductase member 3; aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase)
Gene Aliases: AFAR2; AKR7A3
UniProt ID: (Human) O95154
Entrez Gene ID: (Human) 22977
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support