Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSDVTIVKEG WVQKRGEYIK NWRPRYFLLK TDGSFIGYKE KPQDVDLPYP LNNFSVAKCQ LMKTERPKPN TFIIRCLQWT TVIERTFHVD TPEEREEWTE AIQAVADRLQ RQEEERMNCS PTSQIDNIGE EEMDASTTHH KRKTMNDFDY LKLLGKGTFG KVILVREKAS GKYYAMKILK KEVIIAKDEV AHTLTESRVL KNTRHPFLTS LKYSFQTKDR LCFVMEYVNG GELFFHLSRE RVFSEDRTRF YGAEIVSALD YLHSGKIVYR DLKLENLMLD KDGHIKITDF GLCKEGITDA ATMKTFCGTP EYLAPEVLED NDYGRAVDWW GLGVVMYEMM CGRLPFYNQD HEKLFELILM EDIKFPRTLS SDAKSLLSGL LIKDPNKRLG GGPDDAKEIM RHSFFSGVNW QDVYDKKLVP PFKPQVTSET DTRYFDEEFT AQTITITPPE KYDEDGMDCM DNERRPHFPQ FSYSASGRE
AKT3 (PKB gamma) has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). It is involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, glycogen synthesis and glucose uptake. They are involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, as well as glycogen synthesis and glucose uptake. This kinase has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). Alternatively splice transcript variants encoding distinct isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP434N0250; PKB gamma; Protein kinase Akt-3; Protein kinase B gamma; RAC-gamma serine/threonine protein kinase; RAC-gamma serine/threonine-protein kinase; RAC-PK-gamma; STK-2; v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Gene Aliases: AKT3; MPPH; MPPH2; PKB-GAMMA; PKBG; PRKBG; RAC-gamma; RAC-PK-gamma; STK-2
UniProt ID: (Human) Q9Y243
Entrez Gene ID: (Human) 10000
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support