Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPLCPLAHAM QPQSVLHSGY FHPLLRAWQT ATTTLNASNL IYPIFVTDVP DDIQPITSLP GVARYGVKRL EEMLRPLVEE GLRCVLIFGV PSRVPKDERG SAADSEESPA IEAIHLLRKT FPNLLVACDV CLCPYTSHGH CGLLSENGAF RAEESRQRLA EVALAYAKAG CQVVAPSDMM DGRVEAIKEA LMAHGLGNRV SVMSYSAKFA SCFYGPFRDA AKSSPAFGDR RCYQLPPGAR GLALRAVDRD VREGADMLMV KPGMPYLDIV REVKDKHPDL PLAVYHVSGE FAMLWHGAQA GAFDLKAAVL EAMTAFRRAG ADIIITYYTP QLLQWLKEE
The ALAD enzyme is composed of 8 identical subunits and catalyzes the condensation of 2 molecules of delta-aminolevulinate to form porphobilinogen (a precursor of heme, cytochromes and other hemoproteins). ALAD catalyzes the second step in the porphyrin and heme biosynthetic pathway; zinc is essential for enzymatic activity. ALAD enzymatic activity is inhibited by lead and a defect in the ALAD structural gene can cause increased sensitivity to lead poisoning and acute hepatic porphyria.The ALAD enzyme is composed of 8 identical subunits and catalyzes the condensation of 2 molecules of delta-aminolevulinate to form porphobilinogen (a precursor of heme, cytochromes and other hemoproteins). ALAD catalyzes the second step in the porphyrin and heme biosynthetic pathway; zinc is essential for enzymatic activity. ALAD enzymatic activity is inhibited by lead and a defect in the ALAD structural gene can cause increased sensitivity to lead poisoning and acute hepatic porphyria. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALADH; aminolevulinate, delta-, dehydratase; Delta-aminolevulinic acid dehydratase; Porphobilinogen synthase; testicular tissue protein Li 95
Gene Aliases: ALAD; ALADH; PBGS
UniProt ID: (Human) P13716
Entrez Gene ID: (Human) 210
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support