Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VELEQELAEL HQKDSALLFS SCFVANDSTL FTLAKILPGC EIYSDAGNHA SMIQGIRNSG AAKFVFRHND PDHLKKLLEK SNPKIPKIVA FETVHSMDGA ICPLEELCDV SHQYGALTFV DEVHAVGLYG SRGAGIGERD GIMHKIDIIS GTLGKAFGCV GGYIASTRDL VDMVRSYAAG FIFTTSLPPM VLSGALESVR LLKGEEGQAL RRAHQRNVKH MRQLLMDRGL PVIPCPSHII PIRVGNAALN SKLCDLLLSK HGIYVQAINY PTVPRGEELL RLAPSPHHSP QMMEDFVEKL LLAWTAVGLP LQDVSVAACN FCRRPVHFEL MSEWERSYFG NMGPQYVTTY A
ALAS2 specifies an erythroid-specific mitochondrially located enzyme. It catalyzes the first step in the heme biosynthetic pathway. Defects in this gene cause X-linked pyridoxine-responsive sideroblastic anemia.The product of this gene specifies an erythroid-specific mitochondrially located enzyme. The encoded protein catalyzes the first step in the heme biosynthetic pathway. Defects in this gene cause X-linked pyridoxine-responsive sideroblastic anemia. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-aminolevulinate synthase, erythroid-specific, mitochondrial; 5-aminolevulinic acid synthase 2; ALAS-E; aminolevulinate, delta-, synthase 2; Delta-ALA synthase 2; delta-ALA synthetase; Delta-aminolevulinate synthase 2; PRO2399; RP5-884M20.1
Gene Aliases: ALAS-E; ALAS2; ALASE; ANH1; ASB; SIDBA1; XLDPP; XLEPP; XLSA
UniProt ID: (Human) P22557
Entrez Gene ID: (Human) 212
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support