Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen Sequence: NLNYDTFGLA VPSTLPAGPE IGPSPAPPYG LFVGGRFQAP GARSSRPIRD SSGNLHGYVA EGGAKDIRGA VEAAHQAFPG WAGQSPGARA ALLWALAAAL E
This gene encodes a member of the aldehyde dehydrogenase superfamily. The family members act on aldehyde substrates and use nicotinamide adenine dinucleotide phosphate (NADP) as a cofactor. This gene is conserved in chimpanzee, dog, cow, mouse, rat, and zebrafish. The protein encoded by this gene interacts with maspardin, a protein that when truncated is responsible for Mast syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aldehyde dehydrogenase 16 family, member A1; aldehyde dehydrogenase 16A1; Aldehyde dehydrogenase family 16 member A1
Gene Aliases: 2410004H02Rik; AI452234; ALDH16A1; mFLJ00209
UniProt ID: (Human) Q8IZ83, (Rat) Q3T1L0, (Mouse) Q571I9
Entrez Gene ID: (Human) 126133, (Rat) 361571, (Mouse) 69748
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support