Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RKYMFPMGT PDPEYPDDYS QGGTRLDGKN LVQEWLAKRQ GARYVWNRTE LMQASLDPSV THLMGLFEPG DMKYEIHRDS TLDPSLMEMT EAALRLLSRN PRGFFLFVEG GRIDHGHHES RAYRALTETI MFDDAIERAG QLTSEEDTLS LVTADHSHVF SFGGYPLRGS SIFGLAPGKA RDRKAYTVLL YGNGPGYVLK DGARPDVTES ESGSPEYRQQ SAVPLDGETH AGEDVAVFAR GPQAHLVHGV QEQTFIAHVM AFAACLEPYT ACDLAPRAGT TDAAHPGPSV VPALLPLLAG TLLLLGTATA P (223-532 aa encoded by BC014139)
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The product of this gene is a membrane bound glycosylated enzyme, localized to testis, thymus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alkaline phosphatase 5; Alkaline phosphatase Nagao isozyme; Alkaline phosphatase, germ cell type; Alkaline phosphatase, placental-like; alkaline phosphatase, placental-like 2; ALP-1; EAP; Embryonic alkaline phosphatase; Embryonic-type alkaline phosphatase; GCAP; Germ cell alkaline phosphatase; Nagao isozyme; Placental alkaline phosphatase-like; placental-like alkaline phosphatase; PLAP-like; testicular and thymus alkaline phosphatase
Gene Aliases: Akp5; ALPG; ALPPL; ALPPL2; C77216; D1Ertd816e; EAP; GCAP; RGD1565100
UniProt ID: (Human) P10696, (Mouse) P24823
Entrez Gene ID: (Human) 251, (Rat) 367308, (Mouse) 11650
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support