Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TVTVGPGAS GVCPTACICA TDIVSCTNKN LSKVPGNLFR LIKRLDLSYN RIGLLDSEWI PVSFAKLNTL ILRHNNITSI STGSFSTTPN LKCLDLSSNK LKTVKNAVFQ ELKVLEVLLL YNNHISYLDP SAFGGLSQLQ KLYLSGNFLT QFPMDLYVGR FKLAELMFLD VSYNRIPSMP MHHINLVPGK QLRGIYLHGN PFVCDCSLYS LLVFWYRRHF SSVMDFKNDY TCRLWSDSRH SRQVLLLQDS FMNCSDSIIN GSFRALGFIH EAQVGERLMV HCDSKTGNAN TDFIWVGPDN RLLEPDKEME NFYVFHNGSL VIESPRFEDA GVYSCIAMNK QRLLNETVDV TINVSNFTVS RSHAHEAFNT A (30-399 aa encoded by BC047595)
A novel primary response gene, Alivin 1 (ali1), is an activity-dependent gene and promotes survival of neurons. Sequence analyses reveal that rat, mouse, and human Ali1 proteins contain seven leucine-rich repeats, one IgC2-like loop and a transmembrane domain, and display homology to Kek and Trk families. Results suggest that expression of ali1 promotes depolarization-dependent survival of the granule neuron. Mouse ali1 was mapped to a locus approximately 55.3 cM from the centromere on chromosome 15 that is syntenic to positional candidate loci for familial Alzheimer's disease type 5 and Parkinson's disease 8 on human chromosome 12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adhesion molecule with Ig-like domain 2; alivin 1; Alivin-1; AMIGO-2; AMIGO2; amigo2 {ECO:0000312|EMBL:AAO48951.1}; amphoterin induced gene 2; amphoterin induced gene and ORF 2; Amphoterin-induced protein 2; DEGA; differentially expressed in gastric adenocarcinoma; Differentially expressed in gastric adenocarcinomas; transmembrane protein AMIGO2
Gene Aliases: AI415330; ALI1; AMIGO-2; AMIGO2; AW208913; DEGA
UniProt ID: (Human) Q86SJ2, (Mouse) Q80ZD9, (Rat) Q7TNJ4
Entrez Gene ID: (Human) 347902, (Mouse) 105827, (Rat) 300186
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support