Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TAMRVAAKN GHSQIIKLLE KYGASSLNGC SPSPVHTMEQ KPLQSLSSKV QSLTIKSNSS GSTGGGDMQP SLRGLPNGPT HAFSSPSESP DSTVDRQKSS LSNNSLKSSK NSSLRTTSST ATAQTVPIDS FHNLSFTEQI QQHSLPRSRS RQSIVSPSST TQSLGQSHNS PSSEFEWSQV KPSLKSTKAS KGGKSENSAK SGSAGKKAKQ SNSSQPKVLE YEMTQFDRRG PIAKSGTAAP PKQMPAESQC KIMIPSAQQE IGRSQQQFLI HQQSGEQKKR NGIMTNPNYH LQSNQVFLGR VSVPRTMQDR GHQEVLEGYP SSETELSLKQ ALKLQIEGSD PSFNYKKETP L (1080-1429 aa encoded by BC024725)
Involved in the endosome-to-plasma membrane trafficking and recycling of SNX27-retromer-dependent cargo proteins, such as GLUT1 (PubMed:25278552). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 50
Gene Aliases: ANKRD50; KIAA1223
UniProt ID: (Human) Q9ULJ7
Entrez Gene ID: (Human) 57182
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support