Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MSTRYHQAAS DSYLELLKEA TKRDLNLSDE DGMTPTLLAA YHGNLEALEI ICSRGGDPDR CDIWGNTPLH FAASNGHAHC VSFLVNFGAN IFALDNDLQT PLDAAASREQ NECVALLDKA ATAQNIMNPK KVTRLKEQAQ KNARRQIKEC ERLQEKHQNK MAHTYSKEES GTLSSSKGTF SRSSPSNASA PGTFGSLSKG IKDTFKIKFK KNKDTAEQVG KEGRSGQRNV MEVFREEEED SFSGDFKEKL QLSAEEDGSV HHESILNRPG LGSIVFRRNR ISSPEDISDS KREFGFKLPS ELLQRQGASE ADEGAADEEG EENGLKDDLP WDDDEVEWEE DVVDATPLEV FLLSQHLEEF LPIFKREQID LEALLLCSDE DLQSIQMQLG PRKKVLNAIN RRKQVLQQPG QLVDTSL
As part of the intermicrovillar adhesion complex/IMAC plays a role in epithelial brush border differentiation, controlling microvilli organization and length. Plays a role in assembly of the complex (PubMed:26812018). May play a role in cellular response to endoplasmic reticulum stress. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat and SAM domain-containing protein 4B; Harmonin-interacting ankyrin repeat-containing protein; harmonin-interacting ankyrin-repeat containing protein; Harp
Gene Aliases: ANKS4B; HARP
UniProt ID: (Human) Q8N8V4
Entrez Gene ID: (Human) 257629
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support