Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSNSFPYNE QSGGGEATEL GQEATSTISP SGAFGLFSSD LKKNEDLKQM LESNKDSAKL DAMKRIVGMI AKGKNASELF PAVVKNVASK NIEIKKLVYV YLVRYAEEQQ DLALLSISTF QRALKDPNQL IRASALRVLS SIRVPIIVPI MMLAIKEASA DLSPYVRKNA AHAIQKLYSL DPEQKEMLIE VIEKLLKDKS TLVAGSVVMA FEEVCPDRID LIHKNYRKLC NLLVDVEEWG QVVIIHMLTR YARTQFVSPW KEGDELEDNG KNFYESDDDQ KEKTDKKKKP YTMDPDHRLL IRNTKPLLQS RNAAVVMAVA QLYWHISPKS EAGIISKSLV RLLRSNREVQ YIVLQNIATM SIQRKGMFEP YLKSFYVRST DPTMIKTLKL EILTNLANEA NISTLLREFQ TYVKSQDKQF AAATIQTIGR CATNILEVTD TCLNGLVCLL SNRDEIVVAE SVVVIKKLLQ MQPAQHGEII KHMAKLLDSI TVPVARASIL WLIGENCERV PKIAPDVLRK MAKSFTSEDD LVKLQILNLG AKLYLTNSKQ TKLLTQYILN LGKYDQNYDI RDRTRFIRQL IVPNVKSGAL SKYAKKIFLA QKPAPLLESP FKDRDHFQLG TLSHTLNIKA TGYLELSNWP EVAPDPSVRN VEVIELAKEW TPAGKAKQEN SAKKFYSESE EEEDSSDSSS DSESESGSES GEQGESGEEG DSNEDSSEDS SSEQDSESGR ESGLENKRTA KRNSKAKGKS DSEDGEKENE KSKTSDSSND ESSSIEDSSS DSESESEPES ESESRRVTKE KEKKTKQDRT PLTKDVSLLD LDDFNPVSTP VALPTPALSP SLMADLEGLH LSTSSSVISV STPAFVPTKT HVLLHRMSGK GLAAHYFFPR QPCIFGDKMV SIQITLNNTT DRKIENIHIG EKKLPIGMKM HVFNPIDSLE PEGSITVSMG IDFCDSTQTA SFQLCTKDDC FNVNIQPPVG ELLLPVAMSE KDFKKEQGVL TGMNETSAVI IAAPQNFTPS VIFQKVVNVA NVGAVPSGQD NIHRFAAKTV HSGSLMLVTV ELKEGSTAQL IINTEKTVIG SVLLRELKPV LSQG
AP3B1 is a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Structurally, AP3B1 is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in AP3B1 are associated with Hermansky-Pudlak syndrome type 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adaptor protein complex AP-3 subunit beta-1; adaptor related protein complex 3, beta 1 subunit; Adaptor-related protein complex 3 subunit beta-1; adaptor-related protein complex 3, beta 1 subunit; AP-3 complex beta-3A subunit; AP-3 complex subunit beta-1; ap3; Beta-3A-adaptin; Clathrin assembly protein complex 3 beta-1 large chain
Gene Aliases: ADTB3; ADTB3A; AP3B1; HPS; HPS2; PE
UniProt ID: (Human) O00203
Entrez Gene ID: (Human) 8546
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support