Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MIKAILIFNN HGKPRLSKFY QPYSEDTQQQ IIRETFHLVS KRDENVCNFL EGGLLIGGSD NKLIYRHYAT LYFVFCVDSS ESELGILDLI QVFVETLDKC FENVCELDLI FHVDKVHNIL AEMVMGGMVL ETNMNEIVTQ IDAQNKLEKS EAGLAGAPAR AVSAVKNMNL PEIPRNINIG DISIKVPNLP SFK
Part of the AP-3 complex, an adapter-related complex which is not clathrin-associated. The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 3 subunit sigma-1; adaptor related protein complex 3, sigma 1 subunit; Adaptor-related protein complex 3 subunit sigma-1; AP-3 complex subunit sigma-1; AP-3 complex subunit sigma-3A; clathrin adaptor complex AP3, sigma-3A subunit; clathrin-associated/assembly/adapter protein, small 3; Clathrin-associated/assembly/adaptor protein, small 3; clathrin-associated/assembly/adaptor protein, small 3 (22kD); Sigma-3A-adaptin; Sigma-adaptin 3a; Sigma3A-adaptin
Gene Aliases: AP3S1; CLAPS3; Sigma3A
UniProt ID: (Human) Q92572
Entrez Gene ID: (Human) 1176
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support