Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Immunogen sequence: MERQVLLSEP EEAAALYRGL SRQPALSAAC LGPEVTTQYG GQYRTVHTEW TQRDLERMEN IRFCRQYLVF HDGDSVVFAG PAGNSVETRG ELLSRESPSG TMKAVLRKAG GTGPGEEKQF LEVWEKNRKL KSFNLSALEK HGPVYEDDCF GCLSWSHSET HLLYVAEKKR PKAESFFQTK ALDVSASDDE IARLKKPDQA IKGDQFVFYE DWGENMVSKS IPVLCVLDVE SGNISVLEGV PENVSPGQAF WAPGDAGVVF; Positive Samples: SW480, Jurkat, HepG2, Mouse liver, Mouse kidney, Mouse small intestine, Rat bone marrow; Cellular Location: Cytoplasm
APEH is the enzyme acylpeptide hydrolase, which catalyzes the hydrolysis of the terminal acetylated amino acid preferentially from small acetylated peptides. The acetyl amino acid formed by this hydrolase is further processed to acetate and a free amino acid by an aminoacylase. APEH gene is located within the same region of chromosome 3 (3p21) as the aminoacylase gene, and deletions at this locus are also associated with a decrease in aminoacylase activity. The acylpeptide hydrolase is a homotetrameric protein of 300 kDa with each subunit consisting of 732 amino acid residues. The protein can play an important role in destroying oxidatively damaged proteins in living cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AARE; Acyl-peptide hydrolase; Acylamino-acid-releasing enzyme; Acylaminoacyl-peptidase; acylpeptide hydrolase; APH; MGC2178; N-acylaminoacyl peptide hydrolase; N-acylaminoacyl-peptide hydrolase; OPH; Oxidized protein hydrolase
Gene Aliases: AARE; ACPH; APEH; APH; D3F15S2; D3S48E; DNF15S2; OPH
UniProt ID: (Human) P13798, (Mouse) Q8R146, (Rat) P13676
Entrez Gene ID: (Human) 327, (Mouse) 235606, (Rat) 24206
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support