Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LSTKLKTLP DEVLTKEVEE LILTESKKVL EDVTGEEFVL FMKILSGLKS LQTVSGRQQL VELVAEQADL EQTFNPSDPD CVDRLLQCTR QAVPLFSKNV HSTRFVTYFC EQVLPNLGTL TTPVEGLDIQ LEVLKLLAEM SSFCGDMEKL ETNLRKLFDK LLEYMPLPPE EAENGENAGN EEPKLQFSYV ECLLYSFHQL GRKLPDFLTA KLNAEKLKDF KIRLQYFARG LQVYIRQLRL ALQGKTGEAL KTEENKIKVV ALKITNNINV LIKDLFHIPP SYKSTVTLSW KPVQKVEIGQ KRASEDTTSG SPPKKSSAGP KRDARQIYNP PSGKYSSNLG NFNYERSLQG K (155-504 aa encoded by BC017709)
This gene encodes an apoptosis inhibitory protein whose expression prevents apoptosis after growth factor deprivation. This protein suppresses the transcription factor E2F1-induced apoptosis and also interacts with, and negatively regulates Acinus, a nuclear factor involved in apoptotic DNA fragmentation. Its depletion enhances the cytotoxic action of the chemotherapeutic drugs. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AAC-11; Antiapoptosis clone 11 protein; API-5; Apoptosis inhibitor 5; Cell migration-inducing gene 8 protein; Fibroblast growth factor 2-interacting factor; fibroblast growth factor 2-interacting factor 2; FIF; FIF C1; migration-inducing protein MIG8; Protein XAGL
Gene Aliases: AAC-11; AAC11; AI196452; API-5; API5; MIG8
UniProt ID: (Human) Q9BZZ5, (Mouse) O35841
Entrez Gene ID: (Human) 8539, (Mouse) 11800
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support