Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MDNQAERESE AGVGLQRDED DAPLCEDVEL QDGDLSPEEK IFLREFPRLK EDLKGNIDKL RALADDIDKT HKKFTK
This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ApoL-VI; apolipoprotein L, 6; Apolipoprotein L-VI; Apolipoprotein L6; DKFZp667M075; FLJ38562; FLJ90164; MGC57495
Gene Aliases: APOL-VI; APOL6; APOLVI; UNQ3095/PRO21341
UniProt ID: (Human) Q9BWW8
Entrez Gene ID: (Human) 80830
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support