Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Antibody detects endogenous levels of total ApoOL.
This gene encodes a protein which contains an apolipoprotein O superfamily domain. This domain is found on proteins in circulating lipoprotein complexes. [provided by RefSeq, Sep 2011].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AAIR8193; Apolipoprotein O-like; family with sequence similarity 121A; MICOS complex subunit MIC27; Protein FAM121A
Gene Aliases: 6720473G16Rik; 9430083G14Rik; APOOL; CXorf33; E130106L15Rik; FAM121A; MIC27; RGD1549725; UNQ8193; UNQ8193/PRO23204
UniProt ID: (Human) Q6UXV4, (Mouse) Q78IK4
Entrez Gene ID: (Human) 139322, (Rat) 317191, (Mouse) 68117
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support