Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEAMSPQQET LGGQPGRSSS LTGVSRLAGG SCTKKKMKTL AERRRSAPSL ILDKALQKRP TTRDSPSASV DTCTFLSSLV CSNRTLLIDG RAELKRGLQR QERHLFLFND LFVVAKIKYN NNFKIKNKIK LTDMWTASCV DEVGEGNTNA MKSFVLGWPT VNFVATFSSP EQKDKWLSLL QRYINLEKEK DYPKSIPLKI FAKDIGNCAY SKTITVMNSD TANEVINMSL PMLGITGSER DYQLWVNSGK EEAPYPLIGH EYPYGIKMSH LRDSALLTPG SKDSTTPFNL QEPFLMEQLP REMQCQFILK PSRLAAAQQL SDSGHKTFKR RRSIINWAFW RGSSTHLDNL PSSPTSPMPG QLFGISLPNI CENDNLPKPV LDMLFFLNQK GPLTKGIFRQ SANVKSCREL KEKLNSGVEV HLDCESIFVI ASVLKDFLRN IPGSIFSSDL YDHWVSVMDQ GNDEEKINTV QRLLDQLPRA NVVLLRYLFG VLHNIEQHSS SNQMTAFNLA VCVAPSILWP PASSSPELEN EFTKKVSLLI QFLIENCLRI FGEEITSLFR EVSVRCDTRE NASDISCFQL NDSSYDSLEN ELNEDVDAPC SDLVKKLGQG SRSMDSVLTL SDYDLDQPEV EGLLTLSDFD LAHSKDEDVQ MKRPLESKPV NILVYTKIPL RDHARAPSAM CTPSYLSTAA ANAAKSLRRH RRCSEPSIDY LDSKLSYLRE FYQKKLRKSS CDAILSQKDE DYLKQNQPLQ EEGKTCFKQS LVTGTDVSKK NATTQNTKKK SLSGSEGNHV KLFPKSKPVA ISVASYSPMS SQDHSKNQPF DVNTSGYSPP HTADALKGPR THRRCSEPNI EDQNRKLTYL RGIYSKKQHK TSCEAGLLHG EEDYLKRHKS LQMEGQKLIN QSLVMGIEVG KSSATNQNTE KVLPPRLNLC PRTSYSSLSS PGTSPSGSSV SSQDSAFSQI SEHSVFTPTE TSSPIDCTFQ AQRKREDLSP DFSNASHVSG MPGPSSGQAC SRPAYTKKDT MEWHSQMHSV TLHPSTWLRN GVASLKNWSL KKKAKAARPE EEKIASPKGP LEPPPHASGV PEANSLQEEQ KDLPLRAAEG LSPVQSAQRC SSSPFQDSER HCSSPFSLVE SRLKLCMKSH EEIEPGSQSS SGSLPWERAS ASSWTLEDAT SPDSGPTVVC DIEDRYLTKD I
ARHGAP20 is a potent GTPase-activating protein (GAP) and convert Rho-type GTPases into an inactive GDP-bound state. It contains a pleckstrin homology (PH) domain, a Ras-associating domain, a Rho-GAP domain and two Annexin-like (ANXL) repeats. ARHGAP20 is a direct downstream target of Rap1 in the neurite outgrowth while it's over expression leads to inactivation of Rho for promoting the neurite outgrowth in a Rap1-dependent manner. ARHGAP20 is a tumor suppressor gene and is inactivated by deletion in breast cancer and also by chromosomal translocation in B-cell chronic lymphocytic leukemia. Reports suggest that it may take part in rearrangements of the cytoskeleton and cell signaling events that occur during spermatogenesis. It is expressed predominantly in human brain, liver, ovary and spinal cord while low expression is found in lymph nodes and fetal liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RA and RhoGAP domain containing protein; RA and RhoGAP domain-containing protein; RARhoGAP; rho GTPase activating protein 20 variant 2; Rho GTPase-activating protein 20; Rho-type GTPase-activating protein 20; RhoGTPase activating protein
Gene Aliases: ARHGAP20; KIAA1391; RARHOGAP
UniProt ID: (Human) Q9P2F6, (Rat) Q6REY9
Entrez Gene ID: (Human) 57569, (Rat) 367085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support